Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250961_gH
          See other CES2A GE Assays ›
          Gene Symbol:
          Ces2a
          Gene Name:
          carboxylesterase 2A
          Gene Aliases:
          9130231C15Rik, AI266984, Ces6
          Chromosome Location:
          Chr.8: 104734003 - 104741634 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250961_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ces2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190330.1 NP_001177259.1 6 - 7 1030 96
          NM_133960.5 NP_598721.1 7 - 8 1129 96
          GenBank mRNA AK078953.1 - 7 - 8 1125 96
          AK165784.1 - 6 - 7 1020 96
          BC024082.1 - 7 - 8 1095 5100146 96
          BC024491.1 - 7 - 8 1104 4981579 96
          BC024517.1 - 7 - 8 1102 4984109 96
          BC025537.1 - 7 - 8 1115 5309880 96
          BC025812.1 - 7 - 8 1102 5096915 96
          BC026643.1 - 7 - 8 1102 4986124 96
          BC031295.1 - 7 - 8 1095 5100144 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001191.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ces2a

          Gene Ontology Categories:

          Function(s) Process(es)

          protein glycosylation
          hydrolase activity
          carboxylic ester hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800009243.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009243.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009243.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009243.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009243.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009239.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009239.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009239.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009239.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009239.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009249.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009249.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009249.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009249.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009249.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009246.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009246.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009246.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009242.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009242.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009242.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009242.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009242.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009240.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009240.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009240.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009240.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009240.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009245.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009245.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009245.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009245.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009245.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009247.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009247.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009247.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009247.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009247.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009248.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009248.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009248.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009248.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009248.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009241.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009241.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009241.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009241.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009241.mm.1 NM_133960.5 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009244.mm.1 TC0800001191.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009244.mm.1 NM_001190330.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009244.mm.1 NM_001190330.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009244.mm.1 NM_133960.5.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009244.mm.1 NM_133960.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header