Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249435_g1
          See other 1700092K14RIK GE Assays ›
          Gene Symbol:
          1700092K14Rik
          Gene Name:
          RIKEN cDNA 1700092K14 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 114198258 - 114199199 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249435_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700092K14Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045930.1 1 - 2 70 58
          GenBank mRNA AK007042.1 - 1 - 2 71 58
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY707110.1 - 1 - 2 68 Chr. 11 - 114198260 - 114199199 [+] on Build GRCm38 58

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1700025D23 gene 1700025D23Rik -102.9kb Sense Chr. 11 - 114091803 - 114095325 on NCBI Build GRCm38
          Downstream predicted gene 11690 Gm11690 OTTMUSG00000003386 15.7kb Anti-sense Chr. 11 - 114214571 - 114238503 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001867.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P102218 AK007042.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09363275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146564 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146545 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09223372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09613981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09658135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146598 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146542 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146600 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146550 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146556 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09739456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146551 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09692202 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146592 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09277420 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146584 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146547 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09545609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09417441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09713970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28146616 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100017487.mm.1 TC1100001867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017487.mm.1 NR_045930.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017487.mm.1 NR_045930.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017488.mm.1 TC1100001867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017488.mm.1 NR_045930.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017488.mm.1 NR_045930.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017489.mm.1 TC1100001867.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017489.mm.1 NR_045930.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017489.mm.1 NR_045930.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 714942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0