Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01249030_g1
          See other AU023762 GE Assays ›
          Gene Symbol:
          AU023762
          Gene Name:
          expressed sequence AU023762
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 113496746 - 113587152 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01249030_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: AU023762
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040760.1 4 - 5 320 98
          GenBank mRNA AK049838.1 - 4 - 5 321 98
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CX244349.1 - 3 - 4 235 Chr. 9 - 113498037 - 113511455 [-] on Build GRCm38 98
          GenBankmRNA DN175711.1 - 3 - 4 211 Chr. 9 - 113498032 - 113511415 [-] on Build GRCm38 98
          GenBankmRNA CF172416.1 - 4 - 5 306 Chr. 9 - 113498272 - 113611735 [-] on Build GRCm38 98
          GenBankmRNA BB597836.2 - 4 - 5 318 Chr. 9 - 113498264 - 113587150 [-] on Build GRCm38 98

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream predicted gene,
          36367
          Gm36367 -187.0kb Anti-sense Chr. 9 - 113287712 - 113310985 on NCBI Build GRCm38
          Downstream predicted gene,
          36745
          Gm36745 136.8kb Sense Chr. 9 - 113635860 - 113644603 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003157.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874227 AK013171.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 476783 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 443366 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 349947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 193557 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 755643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 428238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 228789 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 545416 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 28621 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242012 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930924 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330841 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 118753 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377120 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 798940 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 734715 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 655240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343122 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 13464 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 429165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 714812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 479228 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 70389 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 55423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142162 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63308 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2146668 AK049838.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06932647 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02034436 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02034463 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06932587 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02034483 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06932620 - -
          Whole Mouse Genome G4122A A_52_P803124 AK049838.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P803124 BB426608.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115997 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06932587 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02034483 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02034436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07791763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08026578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08144929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07713328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07568561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07673618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08106778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08009757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07568559 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08184424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07786760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840873 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840917 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840998 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07550075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07654846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08026579 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07458384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08080811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07572386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08053971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07473097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08080808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07796941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08058915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840924 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07791770 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08053970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07956930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07685211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08053967 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08184426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08053968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26841005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07512272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08080817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26840986 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1443888_PM_at - -
          Mouse Genome 430 2.0 Array 1443888_at - -
          Mouse Genome 430 2.0 Array 1443888_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900026681.mm.1 TC0900003157.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026681.mm.1 NR_040760.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026681.mm.1 NR_040760.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026682.mm.1 TC0900003157.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026682.mm.1 NR_040760.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026682.mm.1 NR_040760.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026680.mm.1 TC0900003157.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900026680.mm.1 NR_040760.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900026680.mm.1 NR_040760.1 Refseq
          Mouse Expression Set 430 1443888_PM_at - -
          Mouse Expression Set 430 1443888_at - -
          Murine Genome U74v2 Set 129036_r_at - -
          Murine Genome U74v2 Set 129036_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1443888_at - -
          Affymetrix HT Mouse Genome 430B Array 1443888_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 317846 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0