Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246605_g1
          See other BLMH GE Assays ›
          Gene Symbol:
          Blmh
          Gene Name:
          bleomycin hydrolase
          Gene Aliases:
          AI035728, Bh, Bmh
          Chromosome Location:
          Chr.11: 76945656 - 76987389 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246605_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Blmh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178645.4 NP_848760.1 1 - 2 242 99
          GenBank mRNA AK029274.1 - 1 - 2 161 99
          AK035228.1 - 1 - 2 116 99
          AK040799.1 - 1 - 2 132 99
          AK049461.1 - 1 - 2 154 99
          AK051441.1 - 1 - 2 141 99
          AK053872.1 - 1 - 2 122 99
          AK150438.1 - 1 - 2 141 99
          AK150896.1 - 1 - 2 122 99
          AK167325.1 - 1 - 2 122 99
          AK197894.1 - 1 - 2 46 99
          AK199579.1 - 1 - 2 46 99
          BC024090.1 - 1 - 2 127 5102275 99
          BC027037.1 - 1 - 2 98 5364667 99
          BC027362.1 - 1 - 2 131 4935208 99
          BC027403.1 - 1 - 2 152 4952098 99
          BY087076.1 - 1 - 2 242 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001109.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK035228.1, AK040799.1, AK051441.1, AK053872.1, AK150438.1, AK167325.1, BC024090.1, BC027037.1, BC027362.1, BC027403.1, NM_178645.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Blmh

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          response to xenobiotic stimulus
          response to toxic substance
          cysteine-type endopeptidase activity
          peptidase activity
          cysteine-type peptidase activity
          hydrolase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100010064.mm.1 TC1100001109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010064.mm.1 NM_178645.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010064.mm.1 NM_178645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010063.mm.1 TC1100001109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010063.mm.1 NM_178645.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010063.mm.1 NM_178645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010068.mm.1 TC1100001109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010068.mm.1 NM_178645.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010068.mm.1 NM_178645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010056.mm.1 TC1100001109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010056.mm.1 NM_178645.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010056.mm.1 NM_178645.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010061.mm.1 TC1100001109.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100010061.mm.1 NM_178645.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100010061.mm.1 NM_178645.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 447645 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header