Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01246225_m1
          See other TLK2 GE Assays ›
          Gene Symbol:
          Tlk2
          Gene Name:
          tousled-like kinase 2 (Arabidopsis)
          Gene Aliases:
          4933403M19Rik, PKU-alpha, PKUalpha, Tlk
          Chromosome Location:
          Chr.11: 105167521 - 105283959 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01246225_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tlk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001112705.2 NP_001106176.1 8 - 9 1158 139
          NM_001294329.1 NP_001281258.1 6 - 7 1018 139
          NM_001294331.1 NP_001281260.1 8 - 9 1171 139
          NM_001294334.1 NP_001281263.1 8 - 9 1158 139
          GenBank mRNA AF045252.1 - 8 - 9 1177 139
          AF045253.1 - 7 - 8 605 139
          AF045254.1 - 3 - 4 319 139
          AK014829.1 - 8 - 9 796 139
          AK089925.1 - 6 - 7 546 139
          AK165667.1 - 8 - 9 1161 139
          AX775743.1 - 2 - 3 183 139
          CD554252.1 - 2 - 3 282 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001744.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of chromatin assembly or disassembly
          chromatin organization
          protein phosphorylation
          cellular response to DNA damage stimulus
          cell cycle
          chromosome segregation
          multicellular organism development
          spermatogenesis
          phosphorylation
          peptidyl-serine phosphorylation
          cell differentiation
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          intracellular signal transduction
          cellular response to gamma radiation
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100016478.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016478.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016484.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016484.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016486.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016486.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016497.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016497.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016482.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016482.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016495.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016495.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016485.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016485.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016498.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016498.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016489.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016489.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016479.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016479.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016500.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016500.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016480.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016480.mm.1 NM_001112705.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100016491.mm.1 TC1100001744.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100016491.mm.1 NM_001112705.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0