Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01245907_g1
          See other GRN GE Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01245907_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008175.4 NP_032201.2 2 - 3 363 119
          GenBank mRNA AK013678.1 - 2 - 3 179 119
          AK018744.1 - 2 - 3 204 119
          AK147932.1 - 2 - 3 179 119
          AK149570.1 - 2 - 3 182 119
          AK150125.1 - 2 - 3 178 119
          AK150338.1 - 2 - 3 168 119
          AK150420.1 - 2 - 3 167 119
          AK150717.1 - 2 - 3 179 119
          AK150746.1 - 2 - 3 179 119
          AK150881.1 - 2 - 3 179 119
          AK151007.1 - 2 - 3 179 119
          AK151162.1 - 2 - 3 179 119
          AK151343.1 - 2 - 3 179 119
          AK151713.1 - 2 - 3 179 119
          AK151756.1 - 2 - 3 179 119
          AK152048.1 - 2 - 3 179 119
          AK152087.1 - 2 - 3 179 119
          AK152118.1 - 2 - 3 168 119
          AK152213.1 - 2 - 3 168 119
          AK152705.1 - 2 - 3 179 119
          AK152765.1 - 2 - 3 179 119
          AK152944.1 - 2 - 3 168 119
          AK153183.1 - 2 - 3 168 119
          AK153269.1 - 2 - 3 179 119
          AK153289.1 - 2 - 3 179 119
          AK153418.1 - 2 - 3 179 119
          AK153470.1 - 2 - 3 179 119
          AK154492.1 - 2 - 3 179 119
          AK159397.1 - 2 - 3 168 119
          AK159428.1 - 2 - 3 168 119
          AK159557.1 - 2 - 3 179 119
          AK159809.1 - 2 - 3 179 119
          AK160022.1 - 2 - 3 168 119
          AK160033.1 - 2 - 3 179 119
          AK160074.1 - 2 - 3 168 119
          AK196399.1 - 1 - 2 90 120
          AK205747.1 - 1 - 2 90 119
          BC037047.1 - 1 - 2 160 119
          BC049943.1 - 2 - 3 165 119
          BC129849.1 - 2 - 3 163 40109630 119
          BC129850.1 - 2 - 3 163 40109631 119
          M86736.1 - 2 - 3 161 119
          X62321.1 - 1 - 2 144 119

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001688.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grn

          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          RNA binding
          cytokine activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100015900.mm.1 TC1100001688.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100015900.mm.1 NM_008175.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100015900.mm.1 NM_008175.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 592112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5f68d85665-v9rd4:80/100.66.130.250:80.
          git-commit: a5c360236fb87e3c46c4dd6f38a1a1c611be0521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0