Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244562_g1
          See other 4732419C18RIK GE Assays ›
          Gene Symbol:
          4732419C18Rik
          Gene Name:
          RIKEN cDNA 4732419C18 gene
          Gene Aliases:
          Gm3866
          Chromosome Location:
          Chr.8: 123744381 - 123754223 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244562_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4732419C18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK076318.1 - 2 - 3 307 85
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CK031733.1 - 3 - 4 260 Chr. 8 - 123746993 - 123749191 [-] on Build GRCm38 85
          GenBankmRNA BB612800.1 - 2 - 3 307 Chr. 8 - 123747031 - 123754228 [-] on Build GRCm38 85

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 6030466F02 gene 6030466F02Rik -7.1kb Anti-sense Chr. 8 - 123733958 - 123739941 on NCBI Build GRCm38
          Downstream RAB4A,
          member RAS oncogene family
          Rab4a AI848268,
          Rab4
          57.9kb Anti-sense Chr. 8 - 123805847 - 123835291 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702068 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 303491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112371 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58075 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P119350 AK076318.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P119350 AK076318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P293474 AK076318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P119350 AK076318.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P119350 AK076318.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110822 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241411 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06824938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07041016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241359 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07134357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07096351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07003146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241421 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241432 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07274467 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07435825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06824936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241408 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241433 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07403950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07101582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241424 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06824941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241419 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07362940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241428 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241354 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241426 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241434 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241422 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07064111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07274466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07188241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241363 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07192413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06824939 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1444932_PM_at - -
          Mouse Genome 430 2.0 Array 1444932_PM_at - -
          Mouse Genome 430 2.0 Array 1444932_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042484_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100042484_PM_at - -
          Mouse Expression Set 430 1444932_PM_at - -
          Mouse Expression Set 430 1444932_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 644150 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0