Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01244532_m1
          See other 4933400L20RIK GE Assays ›
          Gene Symbol:
          4933400L20Rik
          Gene Name:
          RIKEN cDNA 4933400L20 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 103603799 - 103680382 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01244532_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4933400L20Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_045730.1 4 - 5 613 65
          GenBank mRNA AK016594.1 - 4 - 5 614 65

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream RIKEN cDNA 1600027J07 gene 1600027J07Rik -317.1kb Anti-sense Chr. 8 - 103343878 - 103347534 on NCBI Build GRCm38
          Downstream predicted gene,
          29682
          Gm29682 408.5kb Anti-sense Chr. 8 - 104088904 - 104110267 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001174.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 764079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 702808 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 541407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 492642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 796679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 452648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 499165 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 622350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 924551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 723606 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344745 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 726679 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 114773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809577 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37673 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7873 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 63140 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 65667 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444211 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01689032 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01815134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444161 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01815149 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01815158 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444190 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06444086 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01689039 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140657 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01815149 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06444086 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01689039 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06444211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124286 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07065443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07065446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06855875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06745384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06676060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07055650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26123974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07140660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07330767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26124055 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800009120.mm.1 TC0800001174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009120.mm.1 NR_045730.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009120.mm.1 NR_045730.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009126.mm.1 TC0800001174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009126.mm.1 NR_045730.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009126.mm.1 NR_045730.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009122.mm.1 TC0800001174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009122.mm.1 NR_045730.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009122.mm.1 NR_045730.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009123.mm.1 TC0800001174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009123.mm.1 NR_045730.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009123.mm.1 NR_045730.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009127.mm.1 TC0800001174.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009127.mm.1 NR_045730.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009127.mm.1 NR_045730.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 804564 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0