Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01243122_g1
          See other 2810433D01RIK GE Assays ›
          Gene Symbol:
          2810433D01Rik
          Gene Name:
          RIKEN cDNA 2810433D01 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 102619507 - 102624416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01243122_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 2810433D01Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_033474.1 2 - 3 289 59
          GenBank mRNA AK019272.1 - 2 - 3 220 59
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY335588.1 - 2 - 3 290 Chr. 11 - 102621508 - 102624416 [-] on Build GRCm38 59
          GenBankmRNA AW012071.1 - 2 - 3 239 Chr. 11 - 102621318 - 102624366 [-] on Build GRCm38 59
          GenBankmRNA BI688461.1 - 2 - 3 213 Chr. 11 - 102620281 - 102624328 [-] on Build GRCm38 59
          GenBankmRNA AA185245.1 - 2 - 3 202 Chr. 11 - 102620202 - 102624328 [-] on Build GRCm38 59
          GenBankmRNA AI592702.1 - 2 - 3 201 Chr. 11 - 102620239 - 102624328 [-] on Build GRCm38 59

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream frizzled homolog 2 (Drosophila) Fzd2 AL033370,
          AW456835,
          Fz10,
          Fzd10,
          Mfz10,
          Mfz10a
          -13.4kb Anti-sense Chr. 11 - 102604431 - 102608058 on NCBI Build GRCm38
          Downstream predicted gene 1564 Gm1564 Gm663 42.8kb Anti-sense Chr. 11 - 102664966 - 102682240 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003849.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 431277 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133740 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105759 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07469939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080587 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09239741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09290456 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09741945 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080554 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010088 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080560 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09579855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080578 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09579856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080591 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080537 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09162274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080572 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09010091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09068296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080599 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09579849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28080540 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1445896_PM_at - -
          Mouse Genome 430 2.0 Array 1445896_at - -
          Mouse Genome 430 2.0 Array 1445896_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77132_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 77132_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100035874.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035874.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035874.mm.1 NR_033474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035878.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035878.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035878.mm.1 NR_033474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035875.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035875.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035875.mm.1 NR_033474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035877.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035877.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035877.mm.1 NR_033474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035879.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035879.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035879.mm.1 NR_033474.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035880.mm.1 TC1100003849.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100035880.mm.1 NR_033474.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100035880.mm.1 NR_033474.1 Refseq
          Mouse Expression Set 430 1445896_PM_at - -
          Mouse Expression Set 430 1445896_at - -
          Affymetrix HT Mouse Genome 430B Array 1445896_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1445896_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107040440 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0