Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100030128.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030128.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030128.mm.1 |
NM_144825.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030127.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030127.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030127.mm.1 |
NM_144825.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030124.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030124.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030124.mm.1 |
NM_144825.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030123.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030123.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030123.mm.1 |
NM_144825.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030125.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030125.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030125.mm.1 |
NM_144825.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030126.mm.1 |
TC1100003269.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100030126.mm.1 |
NM_144825.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100030126.mm.1 |
NM_144825.2 |
Refseq |