Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159551.1 | NP_001153023.1 | 4 - 5 | 572 | 82 | |
NM_010376.4 | NP_034506.1 | 4 - 5 | 572 | 82 | ||
GenBank mRNA | AF483214.1 | - | 4 - 5 | 539 | 82 | |
AF515662.1 | - | 4 - 5 | 451 | 82 | ||
AJ345032.1 | - | 4 - 5 | 582 | 82 | ||
AK007664.1 | - | 4 - 5 | 554 | 82 | ||
AK012600.1 | - | 4 - 5 | 565 | 82 | ||
AK028109.1 | - | 4 - 5 | 541 | 82 | ||
AK049101.1 | - | 4 - 5 | 546 | 82 | ||
AK050664.1 | - | 4 - 5 | 515 | 82 | ||
AK080966.1 | - | 4 - 5 | 534 | 82 | ||
AK159176.1 | - | 4 - 5 | 546 | 82 | ||
AK169922.1 | - | 4 - 5 | 529 | 82 | ||
AK171790.1 | - | 4 - 5 | 500 | 82 | ||
DQ168449.1 | - | 4 - 5 | 588 | 82 |
Set Membership: |
![]() ![]() ![]() ![]() |