Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167884.1 | NP_001161356.1 | 8 - 9 | 1380 | 145 | |
NM_001167885.1 | NP_001161357.1 | 5 - 6 | 1078 | 145 | ||
NM_001167887.1 | NP_001161359.1 | 4 - 5 | 1018 | 145 | ||
NM_001167888.1 | NP_001161360.1 | 5 - 6 | 1078 | 145 | ||
NM_001167889.1 | NP_001161361.1 | 4 - 5 | 1018 | 145 | ||
GenBank mRNA | AK054093.1 | - | 3 - 4 | 614 | 145 | |
AK082660.1 | - | 3 - 4 | 698 | 145 | ||
AK087267.1 | - | 8 - 9 | 1282 | 145 | ||
AK139255.1 | - | 3 - 4 | 583 | 145 | ||
AK152179.1 | - | 4 - 5 | 622 | 145 | ||
AY555192.1 | - | 3 - 4 | 708 | 145 | ||
BC011214.1 | - | 4 - 5 | 644 | 145 |
Set Membership: |
![]() ![]() ![]() ![]() |