Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01236121_g1
          See other TRPT1 GE Assays ›
          Gene Symbol:
          Trpt1
          Gene Name:
          tRNA phosphotransferase 1
          Gene Aliases:
          AU079016, AW045591, Tpt1, Tpt1h
          Chromosome Location:
          Chr.19: 6996131 - 6999048 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01236121_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Trpt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153597.2 NP_705825.2 6 - 7 993 57
          NR_027351.1 5 - 6 823 57
          GenBank mRNA AK038830.1 - 5 - 6 825 57
          AK090183.1 - 5 - 6 2124 57
          AK141582.1 - 780 57
          BC027575.1 - 6 - 7 791 1364225 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000128.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Trpt1

          Gene Ontology Categories:

          Function(s) Process(es)

          tRNA splicing, via endonucleolytic cleavage and ligation
          tRNA processing
          regulation of protein kinase activity
          tRNA 2'-phosphotransferase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 BC027575.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK141582.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK048558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 AK090183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1960641 NM_153597.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 394133 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461183 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 BC027575.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK141582.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK048558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 AK090183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1960641 NM_153597.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 BC027575.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK141582.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK048558.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 AK090183.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P199554 NM_153597.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31937238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148301 - -
          Whole Mouse Genome G4122A A_51_P337883 AK048558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 BC027575.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK141582.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK048558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 AK090183.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P199554 NM_153597.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128150 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14306813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31937232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937240 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14306818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31937241 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10461031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461035 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461036 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461037 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461034 - -
          Affymetrix Mouse Gene 1.0 ST Array 10461029 - -
          Affymetrix HT MG-430 PM Array Plate 1452615_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10461028 - -
          Mouse Genome 430 2.0 Array 1452615_PM_s_at - -
          Mouse Genome 430 2.0 Array 1452615_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107328_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 107328_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900001732.mm.1 TC1900000128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001732.mm.1 NM_153597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001732.mm.1 NM_153597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001732.mm.1 NR_027351.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001732.mm.1 NR_027351.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001731.mm.1 TC1900000128.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900001731.mm.1 NM_153597.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001731.mm.1 NM_153597.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001731.mm.1 NR_027351.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900001731.mm.1 NR_027351.1 Refseq
          Mouse Expression Set 430 1452615_s_at - -
          Mouse Expression Set 430 1452615_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452615_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1452615_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452615_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1452615_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 378453 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5910093 - -
          Illumina mouse-6 v1.0 expression beadchip 5910093 - -
          Illumina mouse-6 v1.0 expression beadchip 6220056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006220056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005910093 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2439751 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2480638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2480638 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2439751 - -
          mouse-6 v1.1 (Illumina) scl000496.1_7-S - -
          mouse-6 v1.1 (Illumina) scl0107328.6_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 6220056 - -
          Illumina Sentrix mouse-6.v1 bead array 5910093 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000496.1_7-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0107328.6_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6220056 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5910093 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2885982 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2480638 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2885981 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2480638 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2885981 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2885982 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0107328.6_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-wxxj5:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0