Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mettl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207522.2 | NP_997405.2 | 5 - 6 | 1326 | 76 | |
GenBank mRNA | AK144620.1 | - | 5 - 6 | 1318 | 76 | |
AK148535.1 | - | 6 - 7 | 1503 | 76 | ||
BC067068.1 | - | 5 - 6 | 1300 | 30606425 | 76 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |