Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900000949.mm.1 |
TC1900000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000949.mm.1 |
NM_022012.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000949.mm.1 |
NM_022012.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000945.mm.1 |
TC1900000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000945.mm.1 |
NM_022012.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000945.mm.1 |
NM_022012.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000946.mm.1 |
TC1900000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000946.mm.1 |
NM_022012.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000946.mm.1 |
NM_022012.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000948.mm.1 |
TC1900000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000948.mm.1 |
NM_022012.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000948.mm.1 |
NM_022012.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000947.mm.1 |
TC1900000079.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000947.mm.1 |
NM_022012.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000947.mm.1 |
NM_022012.3 |
Refseq |