Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01232889_m1
          See other XIST GE Assays ›
          Gene Symbol:
          Xist
          Gene Name:
          inactive X specific transcripts
          Gene Aliases:
          A430022B11, AI314753
          Chromosome Location:
          Chr.X: 103460373 - 103483233 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01232889_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Xist
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_001463.3 6 - 7 10257 150
          NR_001570.2 6 - 7 10257 150
          GenBank mRNA L04961.1 - 6 - 7 10221 150
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA CA880243.1 - 2 - 3 211 Chr. X - 103467794 - 103469352 [-] on Build GRCm38 150

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream testis specific X-linked gene Tsx AV207184 -38.9kb Anti-sense Chr. X - 103409101 - 103424588 on NCBI Build GRCm38
          Downstream Jpx transcript,
          Xist activator (non-protein coding)
          Jpx 2010000I03Rik,
          2510040I06Rik,
          Enox
          24.6kb Anti-sense Chr. X - 103493576 - 103506425 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00002775.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons

          Gene Ontology Categories:

          Function(s)

          dosage compensation by inactivation of X chromosome
          spongiotrophoblast differentiation

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113537 AK051106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113537 NR_001463.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113537 AK051106.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P113537 NR_001463.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 L04961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 NR_001463.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 NR_001570.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 L04961.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 NR_001463.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P300376 NR_001570.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P147879 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177061 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 L04961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 NR_001463.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 NR_001570.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 L04961.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 NR_001463.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P300376 NR_001570.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145453 AK051106.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145453 L04961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145453 NR_001463.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P145453 AK011511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P113537 AK051106.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P113537 NR_001463.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P113537 AK051106.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P113537 NR_001463.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 L04961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 NR_001463.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 NR_001570.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 L04961.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 NR_001463.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P300376 NR_001570.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113537 AK051106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113537 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113537 AK051106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P113537 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145453 AK051106.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145453 L04961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145453 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P145453 AK011511.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 L04961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 NR_001570.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 L04961.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P300376 NR_001570.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362360 NR_001463.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P362360 AK011511.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129520 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14535450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14535457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573071 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573077 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573106 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573105 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573103 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573124 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14560260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573116 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15045280 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573084 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573045 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14535460 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573085 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573056 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573073 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573113 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573079 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14535451 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573131 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573052 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573049 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573055 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573057 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14880737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573138 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573109 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573134 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573070 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573133 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32573074 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10606179 - -
          Affymetrix HT MG-430 PM Array Plate 1427262_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436936_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1427263_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10606178 - -
          Mouse Genome 430 2.0 Array 1427263_PM_at - -
          Mouse Genome 430 2.0 Array 1427262_PM_at - -
          Mouse Genome 430 2.0 Array 1427262_at - -
          Mouse Genome 430 2.0 Array 1436936_s_at - -
          Mouse Genome 430 2.0 Array 1427263_at - -
          Mouse Genome 430 2.0 Array 1436936_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 213742_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 213742_PM_at - -
          Affymetrix Murine Genome 6500A Array Msa12930_at - -
          Affymetrix Mouse Clariom S Array PSR0X00017507.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017507.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017507.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017507.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017507.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017501.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017501.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017501.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017505.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017505.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017505.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017505.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017505.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017509.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017509.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017509.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017509.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017509.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017506.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017506.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017506.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017506.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017506.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017502.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017502.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017502.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017502.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017502.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017503.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017503.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017503.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017503.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017503.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017504.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017504.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017504.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017504.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017504.mm.1 NR_001570.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017508.mm.1 TC0X00002775.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00017508.mm.1 NR_001463.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017508.mm.1 NR_001463.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017508.mm.1 NR_001570.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00017508.mm.1 NR_001570.2 Refseq
          Mouse Expression Set 430 1427262_PM_at - -
          Mouse Expression Set 430 1436936_PM_s_at - -
          Mouse Expression Set 430 1427263_at - -
          Mouse Expression Set 430 1436936_s_at - -
          Mouse Expression Set 430 1427262_at - -
          Mouse Expression Set 430 1427263_PM_at - -
          Murine Genome U74v2 Set 99126_at - -
          Murine Genome U74v2 Set 99126_PM_at - -
          Mouse Genome 430A 2.0 Array 1436936_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1427262_PM_at - -
          Mouse Genome 430A 2.0 Array 1427262_at - -
          Mouse Genome 430A 2.0 Array 1427263_PM_at - -
          Mouse Genome 430A 2.0 Array 1427263_at - -
          Mouse Genome 430A 2.0 Array 1436936_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1427263_at - -
          Affymetrix HT Mouse Genome 430A Array 1427262_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427263_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1427262_at - -
          Affymetrix HT Mouse Genome 430A Array 1436936_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1436936_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106520068 - -
          Illumina mouse-6 v1.0 expression beadchip 102630541 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242589 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2475156 - -
          mouse-6 v1.1 (Illumina) scl0213742.1_91-S - -
          mouse-6 v1.1 (Illumina) scl00213742.1_141-S - -
          Illumina Sentrix mouse-6.v1 bead array 102630541 - -
          Illumina Sentrix mouse-6.v1 bead array 106520068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102630541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106520068 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3123341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3022688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224091 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2475156 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3022688 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3123341 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0