Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01231180_g1
          See other SUV420H1 GE Assays ›
          Gene Symbol:
          Suv420h1
          Gene Name:
          suppressor of variegation 4-20 homolog 1 (Drosophila)
          Gene Aliases:
          AA117471, C630029K18Rik, KMT5B, Suv4-20h1
          Chromosome Location:
          Chr.19: 3764155 - 3818303 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01231180_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Suv420h1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001167884.1 NP_001161356.1 12 - 13 1896 74
          NM_001167885.1 NP_001161357.1 9 - 10 1594 74
          NM_001167886.1 NP_001161358.1 7 - 8 1465 74
          NM_001167887.1 NP_001161359.1 8 - 9 1534 74
          NM_001167888.1 NP_001161360.1 9 - 10 1594 74
          NM_001167889.1 NP_001161361.1 8 - 9 1534 74
          NM_144871.4 NP_659120.3 8 - 9 1525 74
          GenBank mRNA AK082660.1 - 7 - 8 1214 74
          AK087267.1 - 12 - 13 1798 74
          AK139255.1 - 7 - 8 1099 74
          AK152179.1 - 8 - 9 1138 74
          AY555192.1 - 7 - 8 1224 74
          BC011214.1 - 8 - 9 1160 74
          BC075709.1 - 6 - 7 1011 3661032 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000017.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Suv420h1

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin organization
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          muscle organ development
          histone methylation
          methylation
          histone H4-K20 methylation
          histone H4-K20 trimethylation
          histone lysine methylation
          methyltransferase activity
          transferase activity
          histone-lysine N-methyltransferase activity
          histone methyltransferase activity (H4-K20 specific)

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167885.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167885.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167886.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167886.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167887.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167887.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_001167889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_144871.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000135.mm.1 NM_144871.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167885.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167885.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167886.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167886.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167887.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167887.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167888.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167889.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_001167889.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_144871.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000139.mm.1 NM_144871.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000141.mm.1 TC1900000017.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000141.mm.1 NM_001167884.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000141.mm.1 NM_001167884.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000141.mm.1 NM_001167888.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000141.mm.1 NM_001167888.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5c8b7848b5-62jj6:80/100.66.134.230:80.
          git-commit: 0dc7e3604112be0ca6b734ece48a71661969252a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0