Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Suv420h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001167884.1 | NP_001161356.1 | 11 - 12 | 1727 | 73 | |
NM_001167885.1 | NP_001161357.1 | 8 - 9 | 1425 | 73 | ||
NM_001167886.1 | NP_001161358.1 | 6 - 7 | 1296 | 73 | ||
NM_001167887.1 | NP_001161359.1 | 7 - 8 | 1365 | 73 | ||
NM_001167888.1 | NP_001161360.1 | 8 - 9 | 1425 | 73 | ||
NM_001167889.1 | NP_001161361.1 | 7 - 8 | 1365 | 73 | ||
NM_144871.4 | NP_659120.3 | 7 - 8 | 1356 | 73 | ||
GenBank mRNA | AK082660.1 | - | 6 - 7 | 1045 | 73 | |
AK083227.1 | - | 7 - 8 | 1357 | 73 | ||
AK087267.1 | - | 11 - 12 | 1629 | 73 | ||
AK139255.1 | - | 6 - 7 | 930 | 73 | ||
AK152179.1 | - | 7 - 8 | 969 | 73 | ||
AY555192.1 | - | 6 - 7 | 1055 | 73 | ||
BC011214.1 | - | 7 - 8 | 991 | 73 | ||
BC075709.1 | - | 5 - 6 | 842 | 3661032 | 73 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |