Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
TC1900000017.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167884.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167884.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167885.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167885.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167886.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167886.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167887.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167888.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167887.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167888.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167889.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_001167889.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_144871.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900000126.mm.1 |
NM_144871.4 |
Refseq |