Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01218663_m1
          See other 8430436N08RIK GE Assays ›
          Gene Symbol:
          8430436N08Rik
          Gene Name:
          RIKEN cDNA 8430436N08 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 7560688 - 7573801 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01218663_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 8430436N08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040645.1 2 - 3 189 68
          GenBank mRNA AK018465.1 - 2 - 3 190 68

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream thymocyte selection-associated high mobility group box Tox 1700007F02Rik -573.1kb Anti-sense Chr. 4 - 6686353 - 6991552 on NCBI Build GRCm38
          Downstream carbonic anhydrase 8 Car8 AW546993,
          Ca8,
          Cals,
          Cals1,
          Carp,
          wdl
          567.7kb Anti-sense Chr. 4 - 8141493 - 8239041 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000046.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2263877 AK018465.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 522291 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 587287 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2263877 AK018465.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P384051 AK018465.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111540 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03641240 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711209 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711333 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711324 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711233 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03323767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711307 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711328 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711316 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804354 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711227 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03804355 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03849923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711174 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711163 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711178 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711312 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711214 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711169 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711284 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711175 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711320 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711336 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03944918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711211 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711315 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711319 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711302 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711172 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711219 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711318 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711331 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22711217 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1432323_PM_at - -
          Mouse Genome 430 2.0 Array 1432323_PM_at - -
          Mouse Genome 430 2.0 Array 1432323_at - -
          Affymetrix Mouse Clariom S Array PSR0400000303.mm.1 TC0400000046.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400000303.mm.1 NR_040645.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400000303.mm.1 NR_040645.1 Refseq
          Mouse Expression Set 430 1432323_at - -
          Mouse Expression Set 430 1432323_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1432323_at - -
          Affymetrix HT Mouse Genome 430B Array 1432323_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 628668 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0