Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01214706_m1
          See other 1700025M24RIK GE Assays ›
          Gene Symbol:
          1700025M24Rik
          Gene Name:
          RIKEN cDNA 1700025M24 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 73268580 - 73284184 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01214706_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 1700025M24Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040687.1 4 - 5 378 124
          NR_040689.1 2 - 3 206 124
          GenBank mRNA BC048655.1 - 4 - 5 375 124

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream furry homolog-like (Drosophila) Fryl 2010313D22Rik,
          2310004H21Rik,
          2510002A14Rik,
          9030227G01Rik,
          mKIAA0826
          -11.9kb Sense Chr. 5 - 73020191 - 73256645 on NCBI Build GRCm38
          Downstream OCIA domain containing 1 Ociad1 6030432N09Rik,
          AI481327,
          AW557942,
          Asrij,
          B230209J16Rik,
          BB021357,
          Emi2,
          Imi2,
          OCIA,
          TPA018
          21.0kb Anti-sense Chr. 5 - 73292772 - 73314077 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002590.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 517586 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 713983 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60794 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169472 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647889 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 752746 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 412142 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 114275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128805 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05614449 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105383 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05614449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04931998 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796762 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796777 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796757 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04172062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04122103 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796760 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796778 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796775 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04931996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796773 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23796771 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 TC0500002590.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040687.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040687.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040688.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040688.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040689.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500022982.mm.1 NR_040689.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 744456 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101170687 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0