Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00005274.mm.1 |
TC0X00000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005274.mm.1 |
NM_001085544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005274.mm.1 |
NM_001085544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005276.mm.1 |
TC0X00000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005276.mm.1 |
NM_001085544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005276.mm.1 |
NM_001085544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005273.mm.1 |
TC0X00000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005273.mm.1 |
NM_001085544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005273.mm.1 |
NM_001085544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005275.mm.1 |
TC0X00000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005275.mm.1 |
NM_001085544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005275.mm.1 |
NM_001085544.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005277.mm.1 |
TC0X00000779.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00005277.mm.1 |
NM_001085544.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00005277.mm.1 |
NM_001085544.1 |
Refseq |