Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01203541_m1
          See other 4930413E15RIK GE Assays ›
          Gene Symbol:
          4930413E15Rik
          Gene Name:
          RIKEN cDNA 4930413E15 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 118952339 - 118961261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01203541_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930413E15Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_040694.1 4 - 5 1109 195
          GenBank mRNA AK015127.1 - 4 - 5 1110 195

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream mediator complex subunit 13-like Med13l 2210413I17Rik,
          6330591G05Rik,
          9030618F05Rik,
          Thrap2,
          Trap240L,
          mKIAA1025
          -194.5kb Sense Chr. 5 - 118560706 - 118765438 on NCBI Build GRCm38
          Downstream RIKEN cDNA 1700081H04 gene 1700081H04Rik 147.0kb Sense Chr. 5 - 119108236 - 119114543 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500001318.mm.1

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2371301 AK015127.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154727 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728669 - -
          Whole Mouse Genome G4122A A_51_P144624 AK015127.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114458 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029886 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04811346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04789684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029893 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029930 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029885 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04732989 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029891 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24029873 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454128_PM_at - -
          Mouse Genome 430 2.0 Array 1454128_at - -
          Mouse Genome 430 2.0 Array 1454128_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500011522.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011522.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011522.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011518.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011518.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011518.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011520.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011520.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011520.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011523.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011523.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011523.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011525.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011525.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011525.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011519.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011519.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011519.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011521.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011521.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011521.mm.1 NR_040694.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011524.mm.1 TC0500001318.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500011524.mm.1 NR_040694.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500011524.mm.1 NR_040694.1 Refseq
          Mouse Expression Set 430 1454128_at - -
          Mouse Expression Set 430 1454128_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454128_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454128_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 771960 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102340333 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7dfb8678b5-8ph95:80/100.66.134.79:80.
          git-commit: 632cddff25488e9c1007a93c87c1899f725eef0d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0