Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029317.1 | NP_083593.1 | 4 - 5 | 416 | 96 | |
GenBank mRNA | AK006268.1 | - | 4 - 5 | 390 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() |