Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700006747.mm.1 |
TC0700000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006747.mm.1 |
NM_029317.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006747.mm.1 |
NM_029317.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006750.mm.1 |
TC0700000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006750.mm.1 |
NM_029317.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006750.mm.1 |
NM_029317.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006751.mm.1 |
TC0700000814.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700006751.mm.1 |
NM_029317.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700006751.mm.1 |
NM_029317.1 |
Refseq |