Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Izumo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC048526.1 | - | 1 - 2 | 191 | 6772588 | 69 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |