Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009800.4 | NP_033930.1 | 1 - 2 | 377 | 77 | |
GenBank mRNA | AB079601.1 | - | 1 - 2 | 156 | 77 | |
AK140073.1 | - | 1 - 2 | 478 | 77 | ||
AY075026.1 | - | 1 - 2 | 378 | 77 | ||
BC019393.1 | - | 1 - 2 | 362 | 3968862 | 77 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |