Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01201979_m1
          See other GNA12 GE Assays ›
          Gene Symbol:
          Gna12
          Gene Name:
          guanine nucleotide binding protein, alpha 12
          Gene Aliases:
          AI414047, AI504261, Galpha12
          Chromosome Location:
          Chr.5: 140759944 - 140830431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01201979_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gna12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010302.2 NP_034432.1 2 - 3 653 62
          GenBank mRNA AK194048.1 - 1 - 2 51 62
          BC111810.1 - 3 - 4 420 40045663 62
          M63659.1 - 2 - 3 653 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003516.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gna12

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          signal transduction
          G protein-coupled receptor signaling pathway
          adenylate cyclase-modulating G protein-coupled receptor signaling pathway
          small GTPase mediated signal transduction
          Rho protein signal transduction
          regulation of cell shape
          regulation of fibroblast migration
          cell differentiation
          regulation of TOR signaling
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          intracellular signal transduction
          embryonic digit morphogenesis
          nucleotide binding
          G protein-coupled receptor binding
          GTPase activity
          GTP binding
          guanyl nucleotide binding
          G-protein beta/gamma-subunit complex binding
          D5 dopamine receptor binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500032263.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032263.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032263.mm.1 NM_010302.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032266.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032266.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032266.mm.1 NM_010302.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032261.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032261.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032261.mm.1 NM_010302.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032265.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032265.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032265.mm.1 NM_010302.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032264.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032264.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032264.mm.1 NM_010302.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032262.mm.1 TC0500003516.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500032262.mm.1 NM_010302.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500032262.mm.1 NM_010302.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0