Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01199418_m1
          See other GSDMCL-PS GE Assays ›
          Gene Symbol:
          Gsdmcl-ps
          Gene Name:
          gasdermin C-like, pseudogene
          Gene Aliases:
          4933426G20Rik
          Chromosome Location:
          Chr.15: 63914461 - 63925453 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01199418_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gsdmcl-ps
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_029414.1 1387 80
          GenBank mRNA AK016931.1 - 1388 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000469.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2312644 AK016931.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 503121 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161145 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P126664 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P126664 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03053127 - -
          Whole Mouse Genome G4122A A_51_P470475 NR_029414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470475 AK016931.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P126664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30278187 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424433 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424434 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424432 - -
          Affymetrix HT MG-430 PM Array Plate 1454049_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424430 - -
          Affymetrix Mouse Gene 1.1 ST Array 10424424 - -
          Affymetrix Mouse Gene 1.1 ST Array 10424418 - -
          Mouse Genome 430 2.0 Array 1454049_at - -
          Mouse Genome 430 2.0 Array 1454049_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500003304.mm.1 TC1500000469.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500003304.mm.1 NR_029414.1 Refseq
          Mouse Expression Set 430 1432380_PM_s_at - -
          Mouse Expression Set 430 1454049_at - -
          Mouse Expression Set 430 1432380_s_at - -
          Mouse Expression Set 430 1454049_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454049_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1454049_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 303237 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246475 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-858577f6d7-cpd7z:80/100.66.134.75:80.
          git-commit: 8a4c7f5d60a2609f21a9257632c46d3db2b553cc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0