Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01198045_g1
          See other 4930408K08RIK GE Assays ›
          Gene Symbol:
          4930408K08Rik
          Gene Name:
          RIKEN cDNA 4930408K08 gene
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01198045_g1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 4930408K08Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK015112.1 - 1 - 2 256 190

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream family with sequence similarity 102,
          member B
          Fam102b 1600010D10Rik,
          B430201A12Rik
          -51.0kb Sense Chr. 3 - 108970997 - 109027607 on NCBI Build GRCm38
          Downstream solute carrier family 25,
          member 54
          Slc25a54 4930443G12Rik,
          SCaMC-1like
          85.0 Anti-sense Chr. 3 - 109080499 - 109120284 on NCBI Build GRCm38

          Back To Top

          More Information


          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 731169 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17476 - -
          Whole Mouse Genome G4122A A_51_P253388 AK015112.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02526604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02577283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22416092 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1454110_PM_at - -
          Mouse Genome 430 2.0 Array 1454110_at - -
          Mouse Genome 430 2.0 Array 1454110_PM_at - -
          Mouse Expression Set 430 1454110_PM_at - -
          Mouse Expression Set 430 1454110_at - -
          Murine Genome U74v2 Set 169970_i_at - -
          Murine Genome U74v2 Set 169970_PM_i_at - -
          Affymetrix HT Mouse Genome 430B Array 1454110_at - -
          Affymetrix HT Mouse Genome 430B Array 1454110_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 793167 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 101940095 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229279 - -
          Illumina Sentrix mouse-6.v1 bead array 101940095 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101940095 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229279 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0