Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168250.1 | NP_001161722.1 | 7 - 8 | 956 | 113 | |
NM_134092.3 | NP_598853.3 | 7 - 8 | 956 | 113 | ||
GenBank mRNA | AK080036.1 | - | 7 - 8 | 770 | 113 | |
AK088549.1 | - | 7 - 8 | 770 | 113 | ||
AK133855.1 | - | 7 - 8 | 804 | 113 | ||
BC052197.1 | - | 7 - 8 | 956 | 6511752 | 113 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |