Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192808_m1
          See other GM11981 GE Assays ›
          Gene Symbol:
          Gm11981
          Gene Name:
          predicted gene 11981
          Gene Aliases:
          OTTMUSG00000005129
          Chromosome Location:
          Chr.11: 6716001 - 6720037 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01192808_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm11981
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_046025.1 4 - 5 428 97
          GenBank mRNA AK076664.1 - 4 - 5 429 97
          BB614940.1 - 4 - 5 429 97
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA AV260002.2 - 4 - 5 398 Chr. 11 - 6715999 - 6720006 [-] on Build GRCm38 97

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream receptor (calcitonin) activity modifying protein 3 Ramp3 AI850306 -38.5kb Anti-sense Chr. 11 - 6658533 - 6677476 on NCBI Build GRCm38
          Downstream adenylate cyclase 1 Adcy1 AC1,
          D11Bwg1392e,
          I-AC,
          brl,
          mKIAA4070
          346.5kb Anti-sense Chr. 11 - 7063489 - 7178506 on NCBI Build GRCm38

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002185.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P140570 AK076664.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370629 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 86059 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P140570 AK076664.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07246441 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140570 AK076664.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07246441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09193497 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09651842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09692158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260941 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09688281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570005 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09485865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09193499 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09193509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569922 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09692161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569990 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569997 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569959 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09193507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09028672 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09193506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27569991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09692165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09172429 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27570024 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100021320.mm.1 TC1100002185.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100021320.mm.1 NR_046025.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100021320.mm.1 NR_046025.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 619942 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0