Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01192778_m1
          See other GM14781 GE Assays ›
          Gene Symbol:
          Gm14781 Mageb5
          Gene Name:
          predicted gene 14781
          melanoma antigen, family B, 5
          Gene Aliases:
          OTTMUSG00000017976, OTTMUSG000000179761
          1700056A17Rik, mMage-b5
          Chromosome Location:
          Chr.X: 91624249 - 91635671 on Build GRCm38
          Chromosome Location:
          Chr.X: 91779559 - 91782976 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm01192778_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm14781
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001205268.1 NP_001192197.1 1 - 2 105 89
          Gene Symbol: Mageb5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028847.1 NP_083123.1 1 - 2 60 89
          GenBank mRNA AK006807.1 - 1 - 2 103 89
          BC116773.1 - 1 - 2 60 40126092 89
          BC116775.1 - 1 - 2 60 40126094 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0X00000871.mm.1, TC0X00002615.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK006807.1, BC116773.1, BC116775.1, NM_001205268.1, NM_028847.1 transcripts.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006955 BC116773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006955 BC116775.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006955 NM_028847.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2006955 AK006807.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006955 BC116773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006955 BC116775.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006955 NM_028847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2006955 AK006807.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P464808 AK006807.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32527799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32527788 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10605657 - -
          Affymetrix Mouse Gene 1.0 ST Array 10605656 - -
          Affymetrix Mouse Gene 1.1 ST Array 10605654 - -
          Mouse Genome 430 2.0 Array 1453280_at - -
          Mouse Genome 430 2.0 Array 1453280_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00005616.mm.1 TC0X00000871.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005616.mm.1 NM_001205268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005616.mm.1 NM_001205268.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016364.mm.1 TC0X00002615.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016364.mm.1 NM_028847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016364.mm.1 NM_028847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016363.mm.1 TC0X00002615.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00016363.mm.1 NM_028847.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00016363.mm.1 NM_028847.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005615.mm.1 TC0X00000871.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005615.mm.1 NM_001205268.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005615.mm.1 NM_001205268.1 Refseq
          Mouse Expression Set 430 1453280_PM_at - -
          Mouse Expression Set 430 1453280_at - -
          Mouse Expression Set 430 1453280_at - -
          Mouse Expression Set 430 1453280_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 396335 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231935 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231935 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-98ffbb479-9qks8:80/100.66.134.79:80.
          git-commit: cc4aab6ff2cd04db3b50ee2b2ac7bb13b20eb648
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0