Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01190430_m1
          See other EPB4.1L2 GE Assays ›
          Gene Symbol:
          Epb4.1l2
          Gene Name:
          erythrocyte protein band 4.1 like 2
          Gene Aliases:
          4.1G, AW555191, D10Ertd398e, Epb41l2, NBL2
          Chromosome Location:
          Chr.10: 25359798 - 25523519 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01190430_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Epb4.1l2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001199265.1 NP_001186194.1 7 - 8 1325 59
          NM_013511.3 NP_038539.2 7 - 8 1216 59
          GenBank mRNA AB370258.1 - 6 - 7 1127 59
          AB370259.1 - 6 - 7 1127 59
          AB370260.1 - 6 - 7 1127 59
          AB370263.1 - 6 - 7 1127 59
          AF044312.1 - 6 - 7 1127 59
          AJ428901.1 - 4 - 5 412 59
          AJ539142.1 - 6 - 7 1127 59
          AJ539143.1 - 6 - 7 1127 59
          AJ539144.1 - 6 - 7 1127 59
          AJ542537.1 - 6 - 7 1127 59
          AK043148.1 - 6 - 7 1104 59
          AK052475.1 - 7 - 8 1217 59
          AK144088.1 - 7 - 8 1326 59
          AK167434.1 - 7 - 8 1242 59
          AK170275.1 - 7 - 8 1268 59
          BC023220.1 - 7 - 8 1212 59
          BC023768.1 - 7 - 8 1238 59
          BC026708.1 - 7 - 8 1197 59
          BC026714.1 - 6 - 7 809 59
          BC029938.1 - 7 - 8 1354 59
          KF534728.1 - 6 - 7 1127 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000224.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Epb4.1l2

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cell shape
          actin cytoskeleton organization
          cortical actin cytoskeleton organization
          actin binding
          structural molecule activity
          protein binding
          cytoskeletal protein binding
          spectrin binding
          PH domain binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000001607.mm.1 TC1000000224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000001607.mm.1 NM_001199265.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001607.mm.1 NM_001199265.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001607.mm.1 NM_013511.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001607.mm.1 NM_013511.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-nqx77:80/100.66.134.75:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0