Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190885.1 | NP_001177814.1 | 7 - 8 | 966 | 131 | |
NM_001190886.1 | NP_001177815.1 | 6 - 7 | 922 | 131 | ||
NM_001290690.1 | NP_001277619.1 | 6 - 7 | 1121 | 131 | ||
NM_027398.3 | NP_081674.2 | 6 - 7 | 933 | 131 | ||
GenBank mRNA | AB075041.1 | - | 6 - 7 | 563 | 131 | |
AJ278534.1 | - | 5 - 6 | 491 | 131 | ||
AK014313.1 | - | 6 - 7 | 921 | 131 | ||
AK045948.1 | - | 7 - 8 | 967 | 131 | ||
AK046398.1 | - | 7 - 8 | 955 | 131 | ||
AK081845.1 | - | 6 - 7 | 923 | 131 | ||
AY050525.1 | - | 7 - 8 | 571 | 131 | ||
AY050526.1 | - | 6 - 7 | 538 | 131 | ||
BC034241.1 | - | 6 - 7 | 875 | 4503251 | 131 | |
BU702946.1 | - | 5 - 6 | 619 | 130 | ||
DQ148493.1 | - | 6 - 7 | 875 | 131 | ||
DQ148494.1 | - | 6 - 7 | 923 | 131 | ||
DQ148495.1 | - | 7 - 8 | 571 | 131 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |