Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930515G16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003100.1 | 1 | 153 | 64 | ||
GenBank mRNA | AK015796.1 | - | 1 | 154 | 64 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | immunoglobulin kappa chain complex | Igk | kappa | Anti-sense | Chr. 6 - 67555636 - 70726754 on NCBI Build GRCm38 | |
Downstream | immunoglobulin kappa variable 1-135 | Igkv1-135 |
Gm4964, bd2 |
44.0kb | Anti-sense | Chr. 6 - 67609745 - 67610508 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |