Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01187598_g1
          See other RNF215 GE Assays ›
          Gene Symbol:
          Rnf215
          Gene Name:
          ring finger protein 215
          Gene Aliases:
          0610009J22Rik, AW557025, C77903
          Chromosome Location:
          Chr.11: 4135153 - 4141193 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01187598_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf215
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027859.2 NP_082135.2 8 - 9 1228 81
          GenBank mRNA AK002414.1 - 8 - 9 1186 81
          AK047194.1 - 1177 81
          AK154140.1 - 6 - 7 1130 81
          BC103627.1 - 8 - 9 1152 6486672 81
          BC115907.1 - 8 - 9 1175 40105152 81
          BC115908.1 - 8 - 9 1175 40105148 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100000035.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf215

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555851 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555852 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166262 BC103627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166262 BM120893.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166262 BM948157.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166262 AK002414.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2166262 AK154140.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948628 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519420 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 649802 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 107485 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P103287 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166262 BC103627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166262 BM120893.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166262 BM948157.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166262 AK002414.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2166262 AK154140.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27555853 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P103287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07239919 - -
          Whole Mouse Genome G4122A A_51_P208019 BC103627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208019 BM120893.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208019 BM948157.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208019 AK002414.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208019 AK154140.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103268 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P103287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09466787 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09466786 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09466793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09466791 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555852 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555850 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27555851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555947 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27555896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555832 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09352936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09466791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09465126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09589188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555842 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555830 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09352934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09352940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09352930 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555863 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09352935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09105124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555837 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555879 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09105110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27555889 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10373871 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373863 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373867 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373872 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373869 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373865 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373870 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373862 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373868 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373864 - -
          Affymetrix Mouse Gene 1.0 ST Array 10373866 - -
          Affymetrix HT MG-430 PM Array Plate 1452756_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10373861 - -
          Mouse Genome 430 2.0 Array 1452756_PM_at - -
          Mouse Genome 430 2.0 Array 1452756_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71673_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71673_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100000441.mm.1 TC1100000035.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000441.mm.1 NM_027859.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000441.mm.1 NM_027859.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000442.mm.1 TC1100000035.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100000442.mm.1 NM_027859.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100000442.mm.1 NM_027859.2 Refseq
          Affymetrix HT Mouse Genome 430B Array 1452756_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1452756_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4540575 - -
          Illumina mouse-6 v1.0 expression beadchip 4540575 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540575 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2718462 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2718462 - -
          mouse-6 v1.1 (Illumina) scl41892.7.15_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 4540575 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl41892.7.15_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540575 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2718462 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2718462 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header