Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01185787_g1
          See other NKAIN1 GE Assays ›
          Gene Symbol:
          Nkain1
          Gene Name:
          Na+/K+ transporting ATPase interacting 1
          Gene Aliases:
          2610200G18Rik, 2810426C15Rik
          Chromosome Location:
          Chr.4: 130530131 - 130574036 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01185787_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nkain1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025998.3 NP_080274.2 5 - 6 693 83
          GenBank mRNA AK011164.1 - 5 - 6 826 83
          AK011850.1 - 5 - 6 624 83
          AK076029.1 - 2 - 3 181 83
          AK184838.1 - 2 - 3 146 83
          AK187377.1 - 2 - 3 146 83
          AK187454.1 - 2 - 3 146 83
          AK205517.1 - 2 - 3 145 83
          AK206217.1 - 2 - 3 146 83
          AK206243.1 - 2 - 3 146 83
          AK208705.1 - 2 - 3 146 83
          AK208715.1 - 2 - 3 146 83
          AK209134.1 - 2 - 3 146 83
          AK209985.1 - 2 - 3 146 83
          AK210264.1 - 2 - 3 146 83
          AK211065.1 - 2 - 3 145 83
          AK211393.1 - 2 - 3 146 83
          AK211541.1 - 2 - 3 146 83
          AK211571.1 - 2 - 3 150 83
          AK213620.1 - 2 - 3 146 83
          AK214155.1 - 2 - 3 83
          AK215330.1 - 2 - 3 146 83
          AK216110.1 - 2 - 3 146 83
          AK216558.1 - 2 - 3 146 83
          AK216625.1 - 2 - 3 146 83
          AK216635.1 - 2 - 3 146 83
          AK217347.1 - 2 - 3 146 83
          AK218211.1 - 2 - 3 146 83
          AK218646.1 - 2 - 3 146 83
          BC116391.1 - 5 - 6 623 40101865 83
          BC116392.1 - 5 - 6 640 83
          EF058048.2 - 5 - 6 746 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003633.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nkain1

          Gene Ontology Categories:

          Process(es)

          ATPase binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400029743.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029743.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029743.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029739.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029739.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029739.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029732.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029732.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029732.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029735.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029735.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029735.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029733.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029733.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029733.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029736.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029736.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029736.mm.1 NM_025998.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029737.mm.1 TC0400003633.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029737.mm.1 NM_025998.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029737.mm.1 NM_025998.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5f68d85665-v9rd4:80/100.66.130.250:80.
          git-commit: a5c360236fb87e3c46c4dd6f38a1a1c611be0521
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0