Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01184589_g1
          See other BRPF1 GE Assays ›
          Gene Symbol:
          Brpf1
          Gene Name:
          bromodomain and PHD finger containing, 1
          Gene Aliases:
          4833438B11Rik, 4930540D11Rik, Brpf2
          Chromosome Location:
          Chr.6: 113306049 - 113324862 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01184589_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Brpf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001282126.1 NP_001269055.1 5 - 6 2296 66
          NM_001282127.1 NP_001269056.1 5 - 6 2064 66
          NM_001282128.1 NP_001269057.1 5 - 6 2296 66
          NM_030178.2 NP_084454.1 5 - 6 2064 66
          GenBank mRNA AK043372.1 - 5 - 6 2297 66
          AK156375.1 - 5 - 6 2244 66
          BC005647.1 - 3 - 4 336 66
          BC031594.1 - 3 - 4 970 66
          BC046521.1 - 5 - 6 2004 5363697 66
          BC138361.1 - 5 - 6 1888 8861381 66
          BC145256.1 - 5 - 6 2120 9053809 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001245.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Brpf1

          Gene Ontology Categories:

          Function(s)

          histone H3 acetylation
          positive regulation of transcription, DNA-templated

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600009608.mm.1 TC0600001245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009608.mm.1 NM_001282126.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009608.mm.1 NM_001282127.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009608.mm.1 NM_001282128.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009608.mm.1 NM_030178.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009609.mm.1 TC0600001245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009609.mm.1 NM_001282126.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009609.mm.1 NM_001282127.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009609.mm.1 NM_001282128.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009609.mm.1 NM_030178.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009611.mm.1 TC0600001245.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600009611.mm.1 NM_001282126.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009611.mm.1 NM_001282127.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009611.mm.1 NM_001282128.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600009611.mm.1 NM_030178.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0