Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600009603.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009603.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009603.mm.1 |
NM_001282127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009603.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009603.mm.1 |
NM_030178.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009604.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009604.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009604.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009602.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009602.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009602.mm.1 |
NM_001282127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009602.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009602.mm.1 |
NM_030178.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009601.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009601.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009601.mm.1 |
NM_001282127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009601.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009601.mm.1 |
NM_030178.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009598.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009598.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009598.mm.1 |
NM_001282127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009598.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009598.mm.1 |
NM_030178.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009605.mm.1 |
TC0600001245.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600009605.mm.1 |
NM_001282126.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009605.mm.1 |
NM_001282127.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009605.mm.1 |
NM_001282128.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600009605.mm.1 |
NM_030178.2 |
Refseq |