Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181487_m1
          See other CYP4X1 GE Assays ›
          Gene Symbol:
          Cyp4x1
          Gene Name:
          cytochrome P450, family 4, subfamily x, polypeptide 1
          Gene Aliases:
          A230025G20, CYPIVX1, CYP_a, Cyp4a28-ps
          Chromosome Location:
          Chr.4: 115108682 - 115133977 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181487_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4x1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001003947.1 NP_001003947.1 11 - 12 1349 69
          GenBank mRNA AJ786769.1 - 11 - 12 1349 69
          AK038526.1 - 1325 69
          BC113125.1 - 11 - 12 1348 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003311.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp4x1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          monooxygenase activity
          iron ion binding
          oxidoreductase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen
          heme binding
          metal ion binding
          aromatase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114580 AK038526.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114580 BC113125.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114580 NM_001003947.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P114580 AJ786769.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480501 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614319 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646049 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112340 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 612209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 198551 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 539110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563651 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 93990 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832599 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 485171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 80914 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 724883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 574987 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 289146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 138928 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 128485 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115489 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114580 AK038526.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114580 BC113125.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114580 NM_001003947.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P114580 AJ786769.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223635 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223624 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23223617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115489 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05366066 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00964581 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115489 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03833916 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03505046 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03833917 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03833924 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03833919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03505045 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03833925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223504 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223451 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223569 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223498 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23223459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223591 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03888984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815164 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422513 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223607 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03422512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03833912 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03943258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04041837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03988789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815165 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815173 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223580 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03888993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03833919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03505045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03815168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03215662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23223587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515170 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515169 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515172 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515177 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515178 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515171 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515174 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515175 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515173 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515179 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515180 - -
          Affymetrix HT MG-430 PM Array Plate 1441662_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10515168 - -
          Mouse Genome 430 2.0 Array 1441662_PM_at - -
          Mouse Genome 430 2.0 Array 1441662_at - -
          Affymetrix Mouse Clariom S Array PSR0400026369.mm.1 TC0400003311.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400026369.mm.1 NM_001003947.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400026369.mm.1 NM_001003947.1 Refseq
          Mouse Expression Set 430 1441662_PM_at - -
          Mouse Expression Set 430 1441662_at - -
          Affymetrix HT Mouse Genome 430B Array 1441662_at - -
          Affymetrix HT Mouse Genome 430B Array 1441662_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 701894 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163399 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163399 - -
          mouse-6 v1.1 (Illumina) GI_51592064-A - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163399 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163399 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0