Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01181468_gH
          See other CYP4A30B GE Assays ›
          Gene Symbol:
          Cyp4a30b
          Gene Name:
          cytochrome P450, family 4, subfamily a, polypeptide 30b
          Gene Aliases:
          Cyp4a30b-ps, OTTMUSG00000008626
          Chromosome Location:
          Chr.4: 115452491 - 115471418 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01181468_gH
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp4a30b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001100185.1 NP_001093655.1 9 - 10 1212 75

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2072091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397458 NM_001104578.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397458 XM_486573.4 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397458 XM_974719.2 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 10716 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67578 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2072091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224494 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224472 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224495 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23224470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224473 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224516 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224490 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224480 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224469 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224494 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224468 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224506 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224462 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224476 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224470 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224571 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224483 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224472 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224508 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224482 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03887327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224500 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224474 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224492 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224502 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23224464 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400008950.mm.1 TC0400001224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008950.mm.1 NM_001100185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008950.mm.1 NM_001100185.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008951.mm.1 TC0400001224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008951.mm.1 NM_001100185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008951.mm.1 NM_001100185.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008949.mm.1 TC0400001224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008949.mm.1 NM_001100185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008949.mm.1 NM_001100185.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008952.mm.1 TC0400001224.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008952.mm.1 NM_001100185.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008952.mm.1 NM_001100185.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 478272 - -
          ABI Mouse Genome Survey Microarray 701366 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-dd6df6ff8-mz6n9:80/100.66.134.79:80.
          git-commit: 27f230ec5a639ef3e18356d3432ca10a74388c39
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.35.0-2025.08.34-1.0