Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012349.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012349.mm.1 |
NM_001033391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012349.mm.1 |
NM_001033391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001033391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001033391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012337.mm.1 |
NM_001170802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012351.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012351.mm.1 |
NM_001033391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012351.mm.1 |
NM_001033391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001033391.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001033391.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012339.mm.1 |
NM_001170802.1 |
Refseq |