Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012346.mm.1 |
NM_001170802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012342.mm.1 |
NM_001170802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012344.mm.1 |
NM_001170802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012340.mm.1 |
NM_001170802.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
TC1000001691.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170800.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170800.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170801.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170801.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170802.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1000012345.mm.1 |
NM_001170802.1 |
Refseq |