Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01176310_m1
          See other RASGRP4 GE Assays ›
          Gene Symbol:
          Rasgrp4
          Gene Name:
          RAS guanyl releasing protein 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.7: 29134933 - 29153961 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01176310_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasgrp4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001174155.1 NP_001167626.1 6 - 7 805 70
          NM_145149.4 NP_660131.2 6 - 7 805 70
          NR_045676.1 6 - 7 805 70
          GenBank mRNA AF331457.1 - 6 - 7 805 70
          AK049562.1 - 5 - 6 548 70
          AK087348.1 - 6 - 7 758 70
          AK089282.1 - 6 - 7 752 70
          AK154746.1 - 6 - 7 795 70
          AK162667.1 - 6 - 7 806 70
          AK165106.1 - 5 - 6 543 70
          AY040628.1 - 6 - 7 805 70
          AY196476.1 - 6 - 7 665 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000522.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasgrp4

          Gene Ontology Categories:

          Function(s) Process(es)

          small GTPase mediated signal transduction
          cell population proliferation
          response to extracellular stimulus
          cell growth
          cell differentiation
          intracellular signal transduction
          positive regulation of GTPase activity
          positive regulation of Ras protein signal transduction
          guanyl-nucleotide exchange factor activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 TC0700000522.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NM_001174155.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NM_001174155.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NM_145149.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NM_145149.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NR_045676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004425.mm.1 NR_045676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 TC0700000522.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NM_001174155.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NM_001174155.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NM_145149.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NM_145149.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NR_045676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004427.mm.1 NR_045676.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 TC0700000522.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NM_001174155.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NM_001174155.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NM_145149.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NM_145149.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NR_045676.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700004426.mm.1 NR_045676.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-854c5b95bd-957wg:80/100.66.134.75:80.
          git-commit: 47437712f32dbf53ebfc670c484059c2aca19dcb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0