Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01174656_g1
          See other ZFP558 GE Assays ›
          Gene Symbol:
          Zfp558
          Gene Name:
          zinc finger protein 558
          Gene Aliases:
          1700007A21Rik, 4932704I17Rik, Zfp558-ps
          Chromosome Location:
          Chr.9: 18453558 - 18478301 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01174656_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfp558
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028935.1 NP_083211.1 7 - 8 840 81
          GenBank mRNA AK005691.1 - 1 - 2 65 81
          AK016584.1 - 9 - 10 2494 81
          BC048749.1 - 6 - 7 548 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001802.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK005691.1 transcripts.

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56488 AK005691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56488 NM_028935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56488 AK005691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P56488 NM_028935.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173758 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 805404 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 613003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 131432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 597046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 532587 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344662 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 464543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26897 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286282 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 126984 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 34415 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122723 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115970 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56488 AK005691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56488 NM_028935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56488 AK005691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P56488 NM_028935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56488 AK005691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56488 NM_028935.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56488 AK005691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P56488 NM_028935.1 mouse_transcriptome
          Agilent Six_ChIP_02_of_03_v2 A_68_P07845148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26337015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26337013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26337014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08104464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07464150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26337009 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1453912_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430973_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10591164 - -
          Affymetrix Mouse Gene 1.1 ST Array 10591161 - -
          Mouse Genome 430 2.0 Array 1430973_at - -
          Mouse Genome 430 2.0 Array 1453912_at - -
          Mouse Genome 430 2.0 Array 1430973_PM_at - -
          Mouse Genome 430 2.0 Array 1453912_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72230_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72230_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900015170.mm.1 TC0900001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015170.mm.1 NM_028935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015170.mm.1 NM_028935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015172.mm.1 TC0900001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015172.mm.1 NM_028935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015172.mm.1 NM_028935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015169.mm.1 TC0900001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015169.mm.1 NM_028935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015169.mm.1 NM_028935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015171.mm.1 TC0900001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015171.mm.1 NM_028935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015171.mm.1 NM_028935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015173.mm.1 TC0900001802.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900015173.mm.1 NM_028935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900015173.mm.1 NM_028935.1 Refseq
          Mouse Expression Set 430 1430973_at - -
          Mouse Expression Set 430 1430973_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453912_at - -
          Affymetrix HT Mouse Genome 430B Array 1453912_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430973_at - -
          Affymetrix HT Mouse Genome 430B Array 1430973_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 777795 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 106980048 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1259136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228386 - -
          Illumina Sentrix mouse-6.v1 bead array 106980048 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106980048 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228386 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6458bd7976-5pn6j:80/100.66.131.68:80.
          git-commit: 78a20c812a412109e55a4daff1692518312115bc
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/redesigning_search_header