Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930413F20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045883.2 | 3 - 4 | 470 | 91 | ||
GenBank mRNA | AK015129.1 | - | 3 - 4 | 471 | 91 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NIPA-like domain containing 2 | Nipal2 |
9330161F08Rik, Npal2 |
Anti-sense | Chr. 15 - 34572799 - 34679437 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 32970 |
Gm32970 | 65.8kb | Anti-sense | Chr. 15 - 34744970 - 34838442 on NCBI Build GRCm38 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |