Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01164233_m1
          See other GM15155 GE Assays ›
          Gene Symbol:
          Gm15155
          Gene Name:
          predicted gene 15155
          Gene Aliases:
          ENSMUSG00000055109, OTTMUSG00000019350
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01164233_m1
          Size
          Availability Made To Order
          Catalog # 4426961
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm15155
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK028243.1 - 4 - 5 505 88
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA BY712647.1 - 4 - 5 505 Chr. X - 155624779 - 156343166 [+] on Build GRCm38 88

          Adjacent Gene Details

          Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
          Upstream patched domain containing 1 Ptchd1 9630036J22Rik,
          AI848149,
          Gm387
          -677.1kb Anti-sense Chr. X - 155569736 - 155623812 on NCBI Build GRCm38
          Downstream gem (nuclear organelle) associated protein 8 pseudogene 4930503H13Rik 236.2kb Sense Chr. X - 156539483 - 156540117 on NCBI Build GRCm38

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1994739 AK028243.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115742 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P110742 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P139095 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1994739 AK028243.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467123 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467303 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466911 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781396 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466974 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467219 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467291 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467313 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466744 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781172 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466957 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P139095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467527 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781530 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466625 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781062 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467535 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781391 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467069 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467107 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467145 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467447 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466943 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781553 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466764 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467203 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466676 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P110742 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466995 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467154 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467014 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466810 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115742 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466849 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467254 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467024 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03781081 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466655 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467473 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467085 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466877 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467104 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467422 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467134 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467459 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466598 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467331 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466932 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09466921 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09467046 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467447 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03781391 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466974 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466810 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467014 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467527 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467473 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115742 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466655 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466849 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466744 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P110742 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467295 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466943 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467535 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467069 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467254 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P139095 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466932 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P03781081 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467123 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467203 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09467331 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09466911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15187417 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760664 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760748 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760658 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14731464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760642 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760631 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14673949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14997325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760655 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760690 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760633 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760654 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14797169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760862 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760684 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15187424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14703657 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760861 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760708 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760686 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760725 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760666 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15004782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760723 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760675 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760635 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760643 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760772 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760714 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760762 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32760615 - -
          Array Name Probe IDs Targets Target Type
          Murine Genome U74v2 Set 136219_r_at - -
          Murine Genome U74v2 Set 136219_PM_r_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101580008 - -
          Illumina mouse-6 v1.0 expression beadchip 101580008 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1238494 - -
          Illumina Sentrix mouse-6.v1 bead array 101580008 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101580008 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2576800 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-vdnrp:80/100.66.131.110:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0