Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agbl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289656.1 | NP_001276585.1 | 18 - 19 | 3054 | 126 | |
NM_001289657.1 | NP_001276586.1 | 17 - 18 | 3028 | 126 | ||
NM_178630.4 | NP_848745.2 | 17 - 18 | 3039 | 126 | ||
GenBank mRNA | AK029793.1 | - | 18 - 19 | 3055 | 126 | |
AK029798.1 | - | 17 - 18 | 2612 | 126 | ||
DQ867031.1 | - | 17 - 18 | 3040 | 126 | ||
DQ867032.1 | - | 17 - 18 | 3040 | 126 |
Set Membership: |
![]() ![]() ![]() ![]() |