Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01150672_m1
          See other ANXA2 GE Assays ›
          Gene Symbol:
          Anxa2
          Gene Name:
          annexin A2
          Gene Aliases:
          AW215814, Cal1h
          Chromosome Location:
          Chr.9: 69453683 - 69491795 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01150672_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Anxa2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007585.3 NP_031611.1 11 - 12 916 78
          GenBank mRNA AK012563.1 - 12 - 13 918 78
          AK088645.1 - 11 - 12 905 78
          AK146096.1 - 11 - 12 904 78
          AK150586.1 - 11 - 12 904 78
          AK152561.1 - 11 - 12 904 78
          AK160746.1 - 11 - 12 904 78
          AK164507.1 - 11 - 12 909 78
          AK165888.1 - 11 - 12 904 78
          AK167894.1 - 11 - 12 903 78
          AK168083.1 - 11 - 12 904 78
          BC003327.1 - 11 - 12 916 3156634 78
          BC004659.1 - 7 - 8 1812 3497991 78
          BC005763.1 - 11 - 12 890 2655513 78
          D10024.1 - 10 - 11 849 78
          M14044.1 - 10 - 11 849 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000918.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Anxa2

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          membrane raft assembly
          positive regulation of protein phosphorylation
          negative regulation of receptor internalization
          vesicle budding from membrane
          body fluid secretion
          collagen fibril organization
          positive regulation of vesicle fusion
          negative regulation of low-density lipoprotein particle receptor catabolic process
          osteoclast development
          fibrinolysis
          negative regulation of catalytic activity
          positive regulation of fibroblast proliferation
          positive regulation of binding
          protein heterotetramerization
          cellular response to acid chemical
          protein localization to plasma membrane
          negative regulation of receptor binding
          positive regulation of signaling receptor activity
          RNA binding
          phospholipase inhibitor activity
          calcium ion binding
          protein binding
          calcium-dependent phospholipid binding
          phosphatidylinositol-4,5-bisphosphate binding
          cytoskeletal protein binding
          phospholipase A2 inhibitor activity
          signaling receptor activator activity
          S100 protein binding
          calcium-dependent protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900007081.mm.1 TC0900000918.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900007081.mm.1 NM_007585.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900007081.mm.1 NM_007585.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-666774c847-7p7bh:80/100.66.135.138:80.
          git-commit: 4f7bff5b1d64937db6e1726f71785770340a1908
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.34.0-2025.08.32-1.0